Få mere at vide
N-Methyl Metribuzin-d3, TRC
Artikelnummer. 30464502
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30464502 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464502 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320297100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl Metribuzin-d3
CAS 1794754-27-0
Molekylær formel C9H13D3N4OS
Mængde 100 mg
Synonym 6-(1,1-Dimethylethyl)-4-[(methyl-d3)amino]-3-(methylthio)-1,2,4-triazin-5(4H)-one, 6-tert-Butyl-4-[(methyl-d3)amino]-3-(methylthio)-1,2,4-triazin-5(4H)-one, Metribuzin Methylamine-d3
InChI formel InChI=1S/C9H16N4OS/c1-9(2,3)6-7(14)13(10-4)8(15-5)12-11-6/h10H,1-5H3/i4D3
SMIL O=C1C(C(C)(C)C)=NN=C(SC)N1NC([2H])([2H])[2H]
IUPAC navn 6-tert-butyl-3-methylsulfanyl-4-(trideuteriomethylamino)-1,2,4-triazin-5-one
Molekylvægt (g/mol) 231.33
Formel vægt 231.1233