Få mere at vide
N-Methyl-L-tyrosine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Methyl-L-tyrosine |
| CAS | 537-49-5 |
| Molekylær formel | C10 H13 N O3 |
| Mængde | 50 mg |
| Synonym | L-Tyrosine, N-methyl-, Surinamine (6CI), Tyrosine, N-methyl-, L- (7CI,8CI), N-Methyl-L-tyrosine, (+)-N-Methyltyrosine, (2S)-3-(4-Hydroxyphenyl)-2-(methylamino)propanoic acid, (2S)-3-(4-Hydroxyphenyl)-2-(methylazaniumyl)propanoate, Andirine, Andrine, L-(+)-N-Methyltyrosine, L-N-Methyltyrosine, N-Methyltyrosine, Ratanhin, Ratanhine, Surinamin |
| InChI formel | InChI=1S/C10H13NO3/c1-11-9(10(13)14)6-7-2-4-8(12)5-3-7/h2-5,9,11-12H,6H2,1H3,(H,13,14)/t9-/m0/s1 |
| SMIL | CN[C@@H](Cc1ccc(O)cc1)C(=O)O |
| IUPAC navn | (2S)-3-(4-hydroxyphenyl)-2-(methylamino)propanoic acid |
| Molekylvægt (g/mol) | 195.22 |
| Formel vægt | 195.0895 |