missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Methyl-L-alanine, 98%

Artikelnummer. 15446578
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15446578 250 mg 250mg
15436578 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15446578 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H60103.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 3913-67-5 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 3913-67-5
Molekylær formel C4H9NO2
Molekylvægt (g/mol) 103.121
MDL nummer MFCD00037241
InChI nøgle GDFAOVXKHJXLEI-VKHMYHEASA-N
Synonym n-methyl-l-alanine, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine, alanine, n-methyl-, l
PubChem CID 5288725
ChEBI CHEBI:17519
IUPAC navn (2S)-2-(methylamino)propanoic acid
SMIL CC(C(=O)O)NC

Tekniske data

Kemisk navn eller materiale N-Methyl-L-alanine
Beilstein 1720922
Mængde 250 mg
Formel vægt 103.12
Procent renhed 98%

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.