Få mere at vide
N-Methyl Gatifloxacin-d3, TRC
Artikelnummer. 30471259
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471259 10 mg
30471578 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471259 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30750210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216721-34-4
Molekylær formel C20H21D3FN3O4
Molekylvægt (g/mol) 392.44
Synonym 1-Cyclopropyl-7-[3-methyl-4-(methyl-d3)-1-piperazinyl]-6-fluoro-1,4-dihydro-8-methoxy -4-oxo-3-quinolinecarboxylic Acid
SMIL FC1=CC(C(C(C(O)=O)=CN2C3CC3)=O)=C2C(OC)=C1N4CC(C)N(C([2H])([2H])[2H])CC4
Tekniske data
Kemisk navn eller materiale N-Methyl Gatifloxacin-d3
Mængde 10 mg
InChI formel InChI=1S/C20H24FN3O4/c1-11-9-23(7-6-22(11)2)17-15(21)8-13-16(19(17)28-3)24(12-4-5-12)10-14(18(13)25)20(26)27/h8,10-12H,4-7,9H2,1-3H3,(H,26,27)/i2D3