Få mere at vide
N-Methyl Duloxetine-d7, TRC
Artikelnummer. 30473291
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30473291 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473291 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3038825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl Duloxetine-d7
CAS 1217657-97-0
Molekylær formel C19H14D7NOS
Mængde 5 mg
Synonym (S)-N,N-Dimethyl-3-(1-naphthalenyl-d7-oxy)-3-(2-thienyl)propanamine, (gammaS)-N,N-Dimethyl-gamma-(1-naphthalenyl-d7-oxy)-2-thiophenepropanamine
InChI formel InChI=1S/C19H21NOS/c1-20(2)13-12-18(19-11-6-14-22-19)21-17-10-5-8-15-7-3-4-9-16(15)17/h3-11,14,18H,12-13H2,1-2H3/i3D,4D,5D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c2c(OC(CCN(C)C)c3cccs3)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn 3-(2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl)oxy-N,N-dimethyl-3-thiophen-2-ylpropan-1-amine
Molekylvægt (g/mol) 318.48
Formel vægt 318.1783