Få mere at vide
N-Methyl Deoxygalactonojirimycin, TRC
Artikelnummer. 30468731
Skift visning
Mængde:
1 mg
2 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468731 1 mg
30466815 2 mg
30481623 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30468731 Leverandør Toronto Research Chemicals Leverandørnr. TRCM4759201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 141206-22-6
Molekylær formel C7 H15 N O4
Molekylvægt (g/mol) 177.198
Synonym (2R,3S,4R,5S)-2-(Hydroxymethyl)-1-methyl-3,4,5-piperidinetriol
IUPAC navn (2R,3S,4R,5S)-2-(hydroxymethyl)-1-methylpiperidine-3,4,5-triol
SMIL CN1C[C@H](O)[C@@H](O)[C@@H](O)[C@H]1CO
Tekniske data
Kemisk navn eller materiale N-Methyl Deoxygalactonojirimycin
Mængde 1 mg
InChI formel InChI=1S/C7H15NO4/c1-8-2-5(10)7(12)6(11)4(8)3-9/h4-7,9-12H,2-3H2,1H3/t4-,5+,6+,7-/m1/s1
Formel vægt 177.1