Få mere at vide
N-Methyl-D3-pyrrolidine, TRC
Artikelnummer. 30471399
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30471399 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471399 Leverandør Toronto Research Chemicals Leverandørnr. TRCM326212250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-d3-pyrrolidine
CAS 77422-30-1
Molekylær formel C5 2H3 H8 N
Mængde 250 mg
Synonym Pyrrolidine, 1-(methyl-d3)- (9CI), 1-(Trideuteriomethyl)pyrrolidine, 1-Methylpyrrolidine D3 (methyl D3), N-Methyl-D3-pyrrolidine
InChI formel InChI=1S/C5H11N/c1-6-4-2-3-5-6/h2-5H2,1H3/i1D3
SMIL [2H]C([2H])([2H])N1CCCC1
IUPAC navn 1-(trideuteriomethyl)pyrrolidine
Molekylvægt (g/mol) 88.17
Formel vægt 88.108