Få mere at vide
N-Methyl-d3-pyrrole, TRC
Artikelnummer. 30474156
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30474156 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474156 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32604750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-d3-pyrrole
CAS 20687-13-2
Molekylær formel C5 2H3 H4 N
Mængde 50 mg
Synonym 1H-Pyrrole, 1-(methyl-d3)- (9CI), Pyrrole, 1-methyl-d3- (8CI), 1-(Methyl-d3)-1H-pyrrole, 1-Methylpyrrole-d3, N-(Methyl-d3)pyrrole, N-Methylpyrrole-D3 (N-methyl-D3), N-Methyl-d3-pyrrole
InChI formel InChI=1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3/i1D3
SMIL [2H]C([2H])([2H])n1cccc1
IUPAC navn 1-(trideuteriomethyl)pyrrole
Molekylvægt (g/mol) 84.13
Formel vægt 84.0767