Få mere at vide
N-Methyl-d3-pyrrole, CDN
Artikelnummer. 30315960
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315960 0.25 g
30322176 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315960 Leverandør CDN Leverandørnr. CDND58530.25G
Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-d3-pyrrole
CAS 20687-13-2
Molekylær formel C5 2H3 H4 N
Mængde 0.25 g
InChI formel InChI=1 S/C5H7N/c1-6-4-2-3-5-6/h2-5 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])n1cccc1
IUPAC navn 1-(trideuteriomethyl)pyrrole
Molekylvægt (g/mol) 84.13
Formel vægt 84.0767 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity