Få mere at vide
N-Methyl-d3-piperazine-2,2,3,3,5,5,6,6-d8, CDN
Artikelnummer. 30309465
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309465 0.25 g
30322598 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309465 Leverandør CDN Leverandørnr. CDND81330.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methylpiperazine-D11
CAS 1319723-22-2
Molekylær formel C5 D11 H N2
Mængde 0.25 g
Synonym Piperazine-2,2,3,3,5,5,6,6-d8, 1-(methyl-d3)- (ACI), 1-(Methyl-d3)piperazine-2,2,3,3,5,5,6,6-d8 (ACI), N-Methyl-d3-piperazine-2,2,3,3,5,5,6,6-d8, N-Methylpiperazine-d11, N-Trideuteriomethyl-2,2,3,3,5,5,6,6-ocatdeuteriopiperazine
InChI formel InChI=1 S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2
SMIL [2 H]C([2 H])([2 H])N1C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuterio-1-(trideuteriomethyl)piperazine
Molekylvægt (g/mol) 111.23
Formel vægt 111.1691 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity