Få mere at vide
N-Methyl-d3-formamide, CDN
Artikelnummer. 30322058
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322058 1 g -
30322571 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322058 Leverandør CDN Leverandørnr. CDND20201G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-d3-formamide
CAS 87955-92-8
Molekylær formel C2 D3 H2 N O
Mængde 1 g
Synonym Formamide, N-(methyl-d3)- (9 CI), N-Methylformamide-D3 (N-methyl-D3), N-(Methyl-d3)formamide, N-Methylformamide-d3
InChI formel InChI=1 S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)/i1D3
SMIL [2 H]C([2 H])([2 H])NC=O
IUPAC navn N-(trideuteriomethyl)formamide
Molekylvægt (g/mol) 62.086
Formel vægt 62.056 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity