Få mere at vide
N-Methyl-d3-epsilon-caprolactam, CDN
Artikelnummer. 30310186
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30310186 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310186 Leverandør CDN Leverandørnr. CDND41511G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylcaprolactam-D3 (N-methyl-D3)
CAS 203645-60-7
Molekylær formel C7 D3 H10 N O
Mængde 1 g
Synonym 2 H-Azepin-2-one, hexahydro-1-(methyl-d3)- (9 CI), Hexahydro-1-(methyl-d3)-2 H-azepin-2-one, 1-(Methyl-d3)azepan-2-one, 1-(Methyl-d3)caprolactam, 1-(Trideuteriomethyl)hexahydro-2 H-azepin-2-one, N-(Methyl-d3)-6-caprolactam, N-(Methyl-d3)-ε-caprolactam, N-Methylcaprolactam-d3, N-Methylcaprolactam-D3 (N-methyl-D3)
InChI formel InChI=1 S/C7H13NO/c1-8-6-4-2-3-5-7(8)9/h2-6H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])N1CCCCCC1=O
IUPAC navn 1-(trideuteriomethyl)azepan-2-one
Molekylvægt (g/mol) 130.203
Formel vægt 130.119 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity