Få mere at vide
N-Methyl-d3-acetamide, CDN
Artikelnummer. 30316669
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316669 0.5 g
30327147 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316669 Leverandør CDN Leverandørnr. CDND52360.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylacetamide-D3 (N-methyl-D3)
CAS 3669-71-4
Molekylær formel C3 D3 H4 N O
Mængde 0.5 g
Synonym Acetamide, N-(methyl-d3)- (7 CI, 8 CI, 9 CI), N-(Methyl-d3)acetamide, N-Methyl-d3-acetamide, N-Methylacetamide-D3 (N-methyl-D3)
InChI formel InChI=1 S/C3H7NO/c1-3(5)4-2/h1-2H3,(H,4,5)/i2D3
SMIL [2 H]C([2 H])([2 H])NC(=O)C
IUPAC navn N-(trideuteriomethyl)acetamide
Molekylvægt (g/mol) 76.112
Formel vægt 76.072 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity