missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Methyl-D-phenylalanine, 97%

Brand:  Alfa Aesar™ H65675.03

Additional Details : CAS Number : 56564-52-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472599

  • 314.73 DKK / 1g


Estimated Shipment
01-09-2020
Add to basket

Specifications

Chemical Identifiers

56564-52-4
179.219
SCIFESDRCALIIM-SECBINFHSA-N
6427037
CNC(CC1=CC=CC=C1)C(=O)O
C10H13NO2
MFCD00069560
r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
(2R)-2-(methylamino)-3-phenylpropanoic acid

Specifications

N-Methyl-D-phenylalanine
C10H13NO2
3199933
SCIFESDRCALIIM-SECBINFHSA-N
(2R)-2-(methylamino)-3-phenylpropanoic acid
6427037
Solid
1g
56564-52-4
MFCD00069560
r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
CNC(CC1=CC=CC=C1)C(=O)O
179.219
179.22
97%
Videos
SDS Documents