Få mere at vide
N-Methyl-Beta-alanine-d3, TRC
Artikelnummer. 30483840
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30483840 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483840 Leverandør Toronto Research Chemicals Leverandørnr. TRCM27657750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-beta-alanine-d3
CAS 1216493-51-4
Molekylær formel C4H6D3NO2
Mængde 50 mg
Synonym N-Methyl-beta-aminopropionic Acid-d3, 3-(Methylamino)propionic acid-d3
InChI formel InChI=1S/C4H9NO2/c1-5-3-2-4(6)7/h5H,2-3H2,1H3,(H,6,7)/i1D3
SMIL [2H]C([2H])([2H])NCCC(=O)O
IUPAC navn 3-(trideuteriomethylamino)propanoic acid
Molekylvægt (g/mol) 106.14
Formel vægt 106.0822