Få mere at vide
N-Methyl Anabasine, TRC
Artikelnummer. 30469431
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469431 10 mg
30473838 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469431 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28750010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl Anabasine
CAS 2055-12-1
Renhedsgrad noter GC
Molekylær formel C11 H16 N2
Mængde 10 mg
Synonym 1-Methyl-anabasine, 3-(1-Methyl-2-piperidinyl)-pyridine, 1-Methyl-2-(3'-pyridyl)piperidine, 1-Methylanabasine, N'-Methylanabasine, N'-Methylanabasine
InChI formel InChI=1S/C11H16N2/c1-13-8-3-2-6-11(13)10-5-4-7-12-9-10/h4-5,7,9,11H,2-3,6,8H2,1H3
SMIL CN1CCCCC1c2cccnc2
IUPAC navn 3-(1-methylpiperidin-2-yl)pyridine
Molekylvægt (g/mol) 176.258
Formel vægt 176.131
Procent renhed >95