Få mere at vide
N-Methyl Amisulpride, TRC
Artikelnummer. 30007165
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007165 50 mg
30006593 10 mg
30007192 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007165 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28732050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391054-22-0
Molekylær formel C18 H29 N3 O4 S
Molekylvægt (g/mol) 383.51
Synonym N-Methylamisulpride, Benzamide, 4-amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxy-N-methyl-, 4-Amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxy-N-methyl-benzamide, 4-Amino-N-[[(2RS)-1-ethylpyrrolidin-2-yl]methyl]-5-(ethylsulfonyl)-2-methoxy-N-methylbenzamide
IUPAC navn 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxy-N-methylbenzamide
SMIL CCN1CCCC1CN(C)C(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC
Tekniske data
Kemisk navn eller materiale N-Methyl Amisulpride
Mængde 50 mg
InChI formel InChI=1S/C18H29N3O4S/c1-5-21-9-7-8-13(21)12-20(3)18(22)14-10-17(26(23,24)6-2)15(19)11-16(14)25-4/h10-11,13H,5-9,12,19H2,1-4H3
Formel vægt 383.1879