missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-4-pyrimidin-2-ylbenzylamine, 90%, Maybridge™
839.64 DKK - 1047.35 DKK
Chemical Identifiers
CAS | 886851-48-5 |
---|---|
Molecular Formula | C12H13N3 |
Molecular Weight (g/mol) | 199.257 |
MDL Number | MFCD09702382 |
InChI Key | DIWVUAYHEYROLA-UHFFFAOYSA-N |
Synonym | n-methyl-4-pyrimidin-2-ylbenzylamine, n-methyl-1-4-pyrimidin-2-yl phenyl methanamine, methyl 4-pyrimidin-2-yl phenyl methyl amine, n-methyl-4-pyrimidin-2-yl benzylamine, benzenemethanamine,n-methyl-4-2-pyrimidinyl, methyl 4-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-4-2-pyrimidinyl-9ci |
PubChem CID | 24229561 |
IUPAC Name | N-methyl-1-(4-pyrimidin-2-ylphenyl)methanamine |
SMILES | CNCC1=CC=C(C=C1)C2=NC=CC=N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10451514
|
Maybridge
CC56246CB |
250mg | Amber Glass Bottle |
839.64 DKK
250mg |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10002164
|
Maybridge
CC56246DA |
1g | Amber Glass Bottle |
1047.35 DKK
1g |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
886851-48-5 | |
199.257 | |
DIWVUAYHEYROLA-UHFFFAOYSA-N | |
24229561 | |
CNCC1=CC=C(C=C1)C2=NC=CC=N2 |
C12H13N3 | |
MFCD09702382 | |
n-methyl-4-pyrimidin-2-ylbenzylamine, n-methyl-1-4-pyrimidin-2-yl phenyl methanamine, methyl 4-pyrimidin-2-yl phenyl methyl amine, n-methyl-4-pyrimidin-2-yl benzylamine, benzenemethanamine,n-methyl-4-2-pyrimidinyl, methyl 4-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-4-2-pyrimidinyl-9ci | |
N-methyl-1-(4-pyrimidin-2-ylphenyl)methanamine |
Specifications
N-methyl-4-pyrimidin-2-ylbenzylamine | |
250mg | |
89.5 | |
90% | |
MFCD09702382 | |
DIWVUAYHEYROLA-UHFFFAOYSA-N | |
N-methyl-1-(4-pyrimidin-2-ylphenyl)methanamine | |
24229561 | |
90% |
Amber Glass Bottle | |
886851-48-5 | |
100.0 | |
C12H13N3 | |
n-methyl-4-pyrimidin-2-ylbenzylamine, n-methyl-1-4-pyrimidin-2-yl phenyl methanamine, methyl 4-pyrimidin-2-yl phenyl methyl amine, n-methyl-4-pyrimidin-2-yl benzylamine, benzenemethanamine,n-methyl-4-2-pyrimidinyl, methyl 4-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-4-2-pyrimidinyl-9ci | |
CNCC1=CC=C(C=C1)C2=NC=CC=N2 | |
199.257 | |
199.26 |