Få mere at vide
N’-Methyl-4-nitrophenylene-1,2-diamine-d3, TRC
Artikelnummer. 30480389
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480389 50 mg
30473357 500 mg
30472475 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480389 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32533750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246816-48-7
Molekylær formel C7 H6 D3 N3 O2
Molekylvægt (g/mol) 170.18
Synonym N1-Methyl-4-nitro-1,2-benzenediamine, N1-Methyl-4-nitro-o-phenylenediamine, 2-Methylamino-5-nitroaniline, N1-Methyl-4-nitro-1,2-benzenediamine, N1-Methyl-4-nitro-o-phenylenediamine
IUPAC navn 4-nitro-1-N-(trideuteriomethyl)benzene-1,2-diamine
SMIL [2H]C([2H])([2H])Nc1ccc(cc1N)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale N'-Methyl-4-nitrophenylene-1,2-diamine-d3
Mængde 50 mg
InChI formel InChI=1S/C7H9N3O2/c1-9-7-3-2-5(10(11)12)4-6(7)8/h2-4,9H,8H2,1H3/i1D3
Formel vægt 170.0883