Få mere at vide
N-Methyl-4-aminobutyric Acid-d3, TRC
Artikelnummer. 30474766
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30474766 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474766 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2857475MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-4-aminobutyric Acid-d3
CAS 1215511-11-7
Molekylær formel C5H8D3NO2
Mængde 5 mg
Synonym 4-(Trideuteriomethylamino)butanoic acid, 4-(Methyl-d3-amino)butanoic acid, 4-(Methyl-d3-amino)butyric acid, 4-(N-Methylamino)butanoic acid-D3 (N-methyl-D3), N-(Methyl-d3)-4-aminobutyric acid, N-(Methyl-d3)-GABA, N-(Methyl-d3)-γ-aminobutyric acid, N-Methyl-4-aminobutyric acid-d3
InChI formel InChI=1S/C5H11NO2/c1-6-4-2-3-5(7)8/h6H,2-4H2,1H3,(H,7,8)/i1D3
SMIL [2H]C([2H])([2H])NCCCC(=O)O
Molekylvægt (g/mol) 120.16