Få mere at vide
N-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxamide Hydrochloride, TRC
Artikelnummer. 30474930
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474930 50 mg
30475495 100 mg
30474518 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30474930 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33424850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1219957-27-3
Molekylær formel C8H12N4O . HCl
Molekylvægt (g/mol) 216.67
Synonym 1H-Pyrazolo[4,3-c]pyridine-3-carboxamide, 4,5,6,7-tetrahydro-N-methyl-, hydrochloride (1:1) (ACI), N-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride, 4,5,6,7-Tetrahydro-N-methyl-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride, N-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
IUPAC navn N-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide;hydrochloride
SMIL Cl.CNC(=O)c1n[nH]c2CCNCc12
Tekniske data
Kemisk navn eller materiale N-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxamide Hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C8H12N4O.ClH/c1-9-8(13)7-5-4-10-3-2-6(5)11-12-7;/h10H,2-4H2,1H3,(H,9,13)(H,11,12);1H
Formel vægt 216.0778