missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(3-methoxythien-2-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-3881620
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
944450-93-5
C7H11NOS
157.23
MFCD11841072
HDALHOAZSOWZLX-UHFFFAOYSA-N
3-methoxythiophen-2-yl methyl methyl amine, 3-methoxythiophen-2-yl-n-methylmethylamine, n-methyl-3-methoxythien-2-yl methylamine, 3-methoxy-2-methylamino methyl thiophene, 1-3-methoxythiophen-2-yl-n-methylmethanamine, 3-methoxy 2-thienyl methyl methylamine
43811050
1-(3-methoxythiophen-2-yl)-N-methylmethanamine
CNCC1=C(OC)C=CS1
This item is not returnable. View return policy
944450-93-5
C7H11NOS
157.23
MFCD11841072
HDALHOAZSOWZLX-UHFFFAOYSA-N
3-methoxythiophen-2-yl methyl methyl amine, 3-methoxythiophen-2-yl-n-methylmethylamine, n-methyl-3-methoxythien-2-yl methylamine, 3-methoxy-2-methylamino methyl thiophene, 1-3-methoxythiophen-2-yl-n-methylmethanamine, 3-methoxy 2-thienyl methyl methylamine
43811050
1-(3-methoxythiophen-2-yl)-N-methylmethanamine
CNCC1=C(OC)C=CS1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 157.24
Percent Purity 97%
Chemical Name or Material N-methyl-(3-methoxythien-2-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'