Få mere at vide
N-Methyl 3,3-Diphenylpropionamide-d3, TRC
Artikelnummer. 30479954
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30479954 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479954 Leverandør Toronto Research Chemicals Leverandørnr. TRCM303707250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl 3,3-Diphenylpropionamide-d3
CAS 1189967-22-3
Molekylær formel C16H14D3NO
Mængde 250 mg
Synonym N-(Methyl-d3)-beta-phenyl-benzenepropanamide
InChI formel InChI=1S/C16H17NO/c1-17-16(18)12-15(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-11,15H,12H2,1H3,(H,17,18)/i1D3
SMIL [2H]C([2H])([2H])NC(=O)CC(c1ccccc1)c2ccccc2
IUPAC navn 3,3-diphenyl-N-(trideuteriomethyl)propanamide
Molekylvægt (g/mol) 242.33
Formel vægt 242.1498