Få mere at vide
N-Methyl-2-piperidinemethyl-d5 Chloride, TRC
Artikelnummer. 30476999
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30476999 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476999 Leverandør Toronto Research Chemicals Leverandørnr. TRCM324812250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-2-piperidinemethyl-d5 Chloride
CAS 1795025-62-5
Molekylær formel C7H9D5ClN
Mængde 250 mg
Synonym 1-Methyl-2-chloromethylpiperidine-d5, 2-Chloromethyl-1-methylpiperidine-d5
InChI formel InChI=1S/C7H14ClN/c1-9-5-3-2-4-7(9)6-8/h7H,2-6H2,1H3/i1D3,6D2
SMIL [2H]C([2H])([2H])N1CCCCC1C([2H])([2H])Cl
IUPAC navn 2-[chloro(dideuterio)methyl]-1-(trideuteriomethyl)piperidine
Molekylvægt (g/mol) 152.68
Formel vægt 152.1129