Få mere at vide
N-Methyl-2-phenylethylamine Hydrochloride (N-Methyl-PEA HCl) 1.0 mg/ml in Methanol (as free base), LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Methyl-2-phenylethylamine Hydrochloride |
| CAS | 4104-43-2 |
| Analyt- eller komponentnavne | N-Methyl-2-phenylethylamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H13 N . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | N-Methyl-2-phenylethylamine Hydrochloride, N-Methyl-PEA HCl |
| InChI formel | InChI=1S/C9H13N.ClH/c1-10-8-7-9-5-3-2-4-6-9;/h2-6,10H,7-8H2,1H3;1H |
| Løsningstype | Single Solution |
| SMIL | Cl.CNCCc1ccccc1 |
| Vis mere |