Få mere at vide
N-Methyl-2-nitroaniline-d3, TRC
Artikelnummer. 30466159
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30466159 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466159 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32355750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-2-nitroaniline-d3
CAS 112333-14-9
Molekylær formel C7 2H3 H5 N2 O2
Mængde 50 mg
Synonym Benzenamine','N-(methyl-d3)-2-nitro- (9CI), N-Methyl-2-nitroaniline-d3, N-Methyl-2-nitroaniline-D3 (N-methyl-D3), N-(Methyl-d3)-2-nitrobenzenamine, 2-(Methyl-d3-amino)nitrobenzene, o-Nitro-N-(methyl-d3)aniline
InChI formel InChI=1S/C7H8N2O2/c1-8-6-4-2-3-5-7(6)9(10)11/h2-5,8H,1H3/i1D3
SMIL [2H]C([2H])([2H])Nc1ccccc1[N+](=O)[O-]
IUPAC navn 2-nitro-N-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 155.17
Formel vægt 155.0774