Få mere at vide
N-Methyl-2-[3-[(1RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine Maleate, Mikromol™
Artikelnummer. 17974356
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
17974356 50 mg
1 options
Artikelnummer. 17974356 Leverandør Mikromol Leverandørnr. MM0595.11
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methyl-2-[3-[(1RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine Maleate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-2-[3-[(1RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine Maleate
CAS 135784-57-5
Analyt- eller komponentnavne N-Methyl-2-[3-[(1RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine Maleate
Forsendelsestilstand Room Temperature
Molekylær formel C19 H22 N2 . C4 H4 O4
Certificeringer/compliance ISO 17025
Synonym Dimetindene Maleate Imp. I (EP), Dimetindene Imp. I (EP), N-Methyl-2-[3-[(1RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine Maleate, Dimetindene Maleate Imp. I (EP) as Maleate, Dimetindene Maleate Impurity I as Maleate, Dimetindene Impurity I as Maleate
InChI formel InChI=1S/C19H22N2.C4H4O4/c1-14(18-9-5-6-11-21-18)19-16(10-12-20-2)13-15-7-3-4-8-17(15)19;5-3(6)1-2-4(7)8/h3-9,11,14,20H,10,12-13H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMIL CNCCC1=C(C(C)c2ccccn2)c3ccccc3C1.OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-but-2-enedioic acid;N-methyl-2-[3-(1-pyridin-2-ylethyl)-1H-inden-2-yl]ethanamine
Molekylvægt (g/mol) 394.46
Mængde 50 mg
Formel vægt 394.1893 g/mol