Få mere at vide
N-Methyl-1-(methylthio)-2-nitroethenamine-d3, TRC
Artikelnummer. 30476449
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30476449 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476449 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32302210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-1-(methylthio)-2-nitroethenamine-d3
CAS 1331637-53-6
Molekylær formel C4H5D3N2O2S
Mængde 10 mg
Synonym N-Methyl-1-(methylthio)-2-nitro-ethenamine-d3, 1-(Methylamino)-1-(methylthio)-2-nitroethene-d3, 2-(Methylthio)-2-(methylamino)-1-nitroethene-d3, 1-(Methylthio)-2-nitro-N-methylethyleneamine-d3
InChI formel InChI=1S/C4H8N2O2S/c1-5-4(9-2)3-6(7)8/h3,5H,1-2H3/i1D3
SMIL [2H]C([2H])([2H])NC(=C[N+](=O)[O-])SC
IUPAC navn 1-methylsulfanyl-2-nitro-N-(trideuteriomethyl)ethenamine
Molekylvægt (g/mol) 151.2
Formel vægt 151.0495