missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Methoxycarbonyl Dabigatran Ethyl Ester, TRC

Artikelnummer. 30473053
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473053 1 mg
30466971 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30473053 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2263151MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 211914-96-4
Molekylær formel C29H31N7O5
Molekylvægt (g/mol) 557.6
Synonym N-[[2-[[[4-[Imino[(methoxycarbonyl)amino]methyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-beta-alanine Ethyl Ester, Ethyl 3-(2-(((4-(N-(Methoxycarbonyl)carbamimidoyl)phenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoate
IUPAC navn ethyl 3-[[2-[[4-(N-methoxycarbonylcarbamimidoyl)anilino]methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate
SMIL CCOC(=O)CCN(C(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C(=N)NC(=O)OC)n2C)c4ccccn4

Tekniske data

Kemisk navn eller materiale N-Methoxycarbonyl Dabigatran Ethyl Ester
Mængde 1 mg
InChI formel InChI=1S/C29H31N7O5/c1-4-41-26(37)14-16-36(24-7-5-6-15-31-24)28(38)20-10-13-23-22(17-20)33-25(35(23)2)18-32-21-11-8-19(9-12-21)27(30)34-29(39)40-3/h5-13,15,17,32H,4,14,16,18H2,1-3H3,(H2,30,34,39)
Formel vægt 557.2387
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.