Få mere at vide
N-Lactoyl-tyrosine, TRC
Artikelnummer. 30005312
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30005312 25 mg -
30011078 10 mg -
30008348 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005312 Leverandør Toronto Research Chemicals Leverandørnr. TRCL16532025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1373771-50-6
Molekylær formel C12H15NO5
Molekylvægt (g/mol) 253.25
Synonym ((S)-2-Hydroxypropanoyl)-L-tyrosine
IUPAC navn (2S)-3-(4-hydroxyphenyl)-2-(2-hydroxypropanoylamino)propanoic acid
SMIL CC(O)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Lactoyl-tyrosine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C12H15NO5/c1-7(14)11(16)13-10(12(17)18)6-8-2-4-9(15)5-3-8/h2-5,7,10,14-15H,6H2,1H3,(H,13,16)(H,17,18)/t7?,10-/m0/s1
Mængde 25 mg
Formel vægt 253.095
Procent renhed >95%