Få mere at vide
N-Lactoyl-Phenylalanine, TRC
Artikelnummer. 17954445
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17954445 50 mg
17941535 100 mg
17913345 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17954445 Leverandør Toronto Research Chemicals Leverandørnr. TRCL16531550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 183241-73-8
Molekylær formel C12H15NO4
Molekylvægt (g/mol) 237.25
Synonym ((S)-2-Hydroxypropanoyl)-L-phenylalanine, N-[(2S)-2-Hydroxy-1-oxopropyl]-L-phenylalanine;
IUPAC navn (2S)-2-[[(2S)-2-hydroxypropanoyl]amino]-3-phenylpropanoic acid
SMIL C[C@H](O)C(=O)N[C@@H](Cc1ccccc1)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Lactoyl-Phenylalanine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C12H15NO4/c1-8(14)11(15)13-10(12(16)17)7-9-5-3-2-4-6-9/h2-6,8,10,14H,7H2,1H3,(H,13,15)(H,16,17)/t8-,10-/m0/s1
Mængde 50 mg
Formel vægt 237.1001
Procent renhed >95%