Få mere at vide
N-(L-Prolyl)-Beta-alanine, TRC
Artikelnummer. 30179486
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179486 500 mg
30179201 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179486 Leverandør Toronto Research Chemicals Leverandørnr. TRCP756190500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 112558-45-9
Molekylær formel C8H14N2O3
Molekylvægt (g/mol) 186.21
Synonym N-L-Prolyl-beta-alanine, L-Prolyl-beta-alanine
IUPAC navn 3-[[(2S)-pyrrolidine-2-carbonyl]amino]propanoic acid
SMIL OC(=O)CCNC(=O)[C@@H]1CCCN1
Tekniske data
Kemisk navn eller materiale N-(L-Prolyl)-beta-alanine
InChI formel InChI=1S/C8H14N2O3/c11-7(12)3-5-10-8(13)6-2-1-4-9-6/h6,9H,1-5H2,(H,10,13)(H,11,12)/t6-/m0/s1
Mængde 500 mg
Formel vægt 186.1004