Få mere at vide
N-[(+)-Jasmonoyl]-(L)-isoleucine, TRC
Artikelnummer. 30007700
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007700 25 mg
30005252 50 mg
30003813 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007700 Leverandør Toronto Research Chemicals Leverandørnr. TRCJ21056025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120330-92-9
Molekylær formel C18 H29 N O4
Molekylvægt (g/mol) 323.427
Synonym JA-L-Ile, N-[2-[(1S,2S)-3-Oxo-2-(2Z)-2-penten-1-ylcyclopentyl]acetyl]-L-isoleucine, ent-Jasmonic Acid L-Isoleucine Conjugate, [1S-[1α,2β(Z)]]-N-[[3-Oxo-2-(2-pentenyl)cyclopentyl]acetyl]-L-isoleucine, N-[[(1S,2S)-3-Oxo-2-(2Z)-2-pentenylcyclopentyl]acetyl]-L-isoleucine, N-[2-[(1S,2S)-3-Oxo-2-(2Z)-2-penten-1-ylcyclopentyl]acetyl]-L-isoleucine
IUPAC navn (2S,3S)-3-methyl-2-[[2-[(1S,2S)-3-oxo-2-[(Z)-pent-2-enyl]cyclopentyl]acetyl]amino]pentanoic acid
SMIL CC\C=C/C[C@H]1[C@H](CC(=O)N[C@@H]([C@@H](C)CC)C(=O)O)CCC1=O
Tekniske data
Kemisk navn eller materiale N-[(+)-Jasmonoyl]-(L)-isoleucine
InChI formel InChI=1S/C18H29NO4/c1-4-6-7-8-14-13(9-10-15(14)20)11-16(21)19-17(18(22)23)12(3)5-2/h6-7,12-14,17H,4-5,8-11H2,1-3H3,(H,19,21)(H,22,23)/b7-6-/t12-,13-,14-,17-/m0/s1
Mængde 25 mg
Formel vægt 323.21