missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylbenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7026919
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
102-97-6
C10H15N
149.237
MFCD00008863
LYBKPDDZTNUNNM-UHFFFAOYSA-N
n-isopropylbenzylamine, n-benzylisopropylamine, isopropylbenzylamine, n-benzyl-n-isopropylamine, benzylisopropylamine, benzenemethanamine, n-1-methylethyl, n-benzyl isopropylamine, benzylamine, n-isopropyl, n-1-methylethyl-benzenemethanamine, benzyl propan-2-yl amine
66024
N-benzylpropan-2-amine
CC(C)NCC1=CC=CC=C1
This item is not returnable. View return policy
102-97-6
C10H15N
149.237
MFCD00008863
LYBKPDDZTNUNNM-UHFFFAOYSA-N
n-isopropylbenzylamine, n-benzylisopropylamine, isopropylbenzylamine, n-benzyl-n-isopropylamine, benzylisopropylamine, benzenemethanamine, n-1-methylethyl, n-benzyl isopropylamine, benzylamine, n-isopropyl, n-1-methylethyl-benzenemethanamine, benzyl propan-2-yl amine
66024
N-benzylpropan-2-amine
CC(C)NCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.91
Boiling Point 199°C to 201°C
Flash Point 87°C (190°F)
Refractive Index 1.502
Quantity 100 g
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material N-Isopropylbenzylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'