Få mere at vide
N-Isopropyl Noratropine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Isopropyl Noratropine |
| CAS | 22235-81-0 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C19 H27 N O3 |
| Mængde | 50 mg |
| Synonym | (1R,3r,5S)-8-(1-Methylethyl)-8-aza-bicyclo[3.2.1]oct-3-yl (2RS)-3-hydroxy-2-phenyl-propanoate, 1αH,5αH-Nortropan-3α-ol, 8-isopropyl-, (±)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-(1-methylethyl)-8-azabicyclo[3.2.1]oct-3-yl ester, endo-(±)-, Benzeneacetic acid, α-(hydroxymethyl)-, 8-(1-methylethyl)-8-azabicyclo[3.2.1]oct-3-yl ester, endo-, N-Isopropylnoratropine, Tropic acid, 8-isopropyl-3α-nortropanyl ester, Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-(1-methylethyl)-8-azabicyclo[3.2.1]oct-3-yl ester, Ipratropium Bromide Imp E (EP) |
| InChI formel | InChI=1S/C19H27NO3/c1-13(2)20-15-8-9-16(20)11-17(10-15)23-19(22)18(12-21)14-6-4-3-5-7-14/h3-7,13,15-18,21H,8-12H2,1-2H3/t15-,16+,17+,18-/m0/s1 |
| SMIL | CC(C)N1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3 |
| IUPAC navn | [(1S,5R)-8-propan-2-yl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate |
| Molekylvægt (g/mol) | 317.42 |
| Vis mere |