Få mere at vide
N-Isopropyl-N-methylamine-d7 Hydrochloride, TRC
Artikelnummer. 30408241
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30408241 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408241 Leverandør Toronto Research Chemicals Leverandørnr. TRCI872254250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Isopropyl-N-methylamine-d7 Hydrochloride
CAS 2733969-46-3
Molekylær formel C4H4D7N . (HCl)
Mængde 250 mg
Synonym N-Methylisopropylamine-d7 Hydrochloride, Methylisopropylammonium-d7 Chloride
InChI formel InChI=1S/C4H11N.ClH/c1-4(2)5-3;/h4-5H,1-3H3;1H/i1D3,2D3,4D;
SMIL CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H].Cl
IUPAC navn N-methylpropan-d7-2-amine hydrochloride
Molekylvægt (g/mol) 116.64
Formel vægt 116.11