missing translation for 'onlineSavingsMsg'
Learn More

N-Isobutyryl-2'-deoxyguanosine, 97%, Thermo Scientific™

Catalog Number 15479467
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
142554-22-1
C14H21N5O4
323.35
MFCD02682949
JPFWZGMCLBMEAU-SLOTWNJKNA-N
guanosine,2'-deoxy-n-2-methylpropyl-9ci
67890594
9-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-(2-methylpropylamino)-3H-purin-6-one
CC(C)CNC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
This item is not returnable. View return policy
142554-22-1
C14H21N5O4
323.35
MFCD02682949
JPFWZGMCLBMEAU-SLOTWNJKNA-N
guanosine,2'-deoxy-n-2-methylpropyl-9ci
67890594
9-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-(2-methylpropylamino)-3H-purin-6-one
CC(C)CNC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Isobutyryl-2′-deoxyguanosine, is used as pharmaceutical intermediate.

Solubility
It is water soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information It is water soluble.
Formula Weight 337.34
Percent Purity 97%
Chemical Name or Material N-Isobutyryl-2'-deoxyguanosine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'