missing translation for 'onlineSavingsMsg'
Få mere at vide

N-iso-Propyl-d7-acrylamide, TRC

Artikelnummer. 30180245
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180245 1 mg
30179941 2 mg
30180058 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180245 Leverandør Toronto Research Chemicals Supplier No. TRCI7791241MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219803-32-3
Molekylær formel C6 D7 H4 N O
Molekylvægt (g/mol) 120.2008
Synonym 2-Propenamide, N-[1-(methyl-d3)ethyl-1,2,2,2-d4]- (ACI), N-[1-(Methyl-d3)ethyl-1,2,2,2-d4]-2-propenamide (ACI), N-iso-Propyl-d7-acrylamide, N-(Isopropyl-d7)acrylamide, N-Isopropylacrylamide-d7, N-Isopropylacrylamide-D7 (isoproyl-D7)
IUPAC navn N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)prop-2-enamide
SMIL [2H]C([2H])([2H])C([2H])(NC(=O)C=C)C([2H])([2H])[2H]

Specifications

Kemisk navn eller materiale N-iso-Propyl-d7-acrylamide
Mængde 1 mg
InChI formel InChI=1S/C6H11NO/c1-4-6(8)7-5(2)3/h4-5H,1H2,2-3H3,(H,7,8)/i2D3,3D3,5D
Formel vægt 120.128
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.