Learn More
Alfa Aesar™ N-(Hydroxymethyl)nonanamide, 98%
Click to view available options
Quantity:
10 g
50 g
250 g
Specifications
Melting Point | 83°C to 87°C |
Boiling Point | 83°C to 87°C |
Quantity | 10 g |
Formula Weight | 187.29 |
Percent Purity | 98% |
Chemical Name or Material | N-(Hydroxymethyl)nonanamide |