Få mere at vide
N-Hydroxy Ropinirole Hydrochloride, TRC
Artikelnummer. 30132392
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132392 10 mg
30004713 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132392 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95341010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1542267-72-0
Molekylær formel C16 H24 N2 O2 . Cl H
Molekylvægt (g/mol) 312.83
Synonym 2H-Indol-2-one, 4-[2-(dipropylamino)ethyl]-1,3-dihydro-1-hydroxy-, hydrochloride (1:1)
IUPAC navn 4-[2-(dipropylamino)ethyl]-1-hydroxy-3H-indol-2-one;hydrochloride
SMIL Cl.CCCN(CCC)CCc1cccc2N(O)C(=O)Cc12
Tekniske data
Kemisk navn eller materiale N-Hydroxy Ropinirole Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C16H24N2O2.ClH/c1-3-9-17(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)18(15)20;/h5-7,20H,3-4,8-12H2,1-2H3;1H
Formel vægt 312.1605