Få mere at vide
N-Hydroxy Phentermine Hydrochloride, TRC
Artikelnummer. 30138410
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138410 5 mg
30138173 1 mg
30005071 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138410 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9489905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51835-51-9
Molekylær formel C10H16ClNO
Molekylvægt (g/mol) 201.69
Synonym N-Hydroxy-α,α-dimethylbenzeneethanamine Hydrochloride;
IUPAC navn N-(2-methyl-1-phenylpropan-2-yl)hydroxylamine;hydrochloride
SMIL Cl.CC(C)(Cc1ccccc1)NO
Tekniske data
Kemisk navn eller materiale N-Hydroxy Phentermine Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C10H15NO.ClH/c1-10(2,11-12)8-9-6-4-3-5-7-9;/h3-7,11-12H,8H2,1-2H3;1H
Formel vægt 201.092