Få mere at vide
n-Hexyl-d13 Alcohol, CDN
Artikelnummer. 30321180
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321180 0.5 g 0.5g
30315480 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321180 Leverandør CDN Leverandørnr. CDND39820.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Hexanol-D13
CAS 204244-84-8
Molekylær formel C6 D13 H O
Mængde 0.5 g
Synonym 1-Hexan-1,1,2,2,3,3,4,4,5,5,6,6,6-d13-ol (9 CI), 1-Hexanol-d13, n-Hexyl-d13 alcohol, 1-Hexanol-1,1,2,2,3,3,4,4,5,5,6,6,6-d13
InChI formel InChI=1 S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexan-1-ol
Molekylvægt (g/mol) 115.2549
Formel vægt 115.1861 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity