Få mere at vide
n-Hexyl Acetate-d3, CDN
Artikelnummer. 30321725
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321725 0.5 g 0.5g
30314069 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321725 Leverandør CDN Leverandørnr. CDND72320.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexyl Acetate-D3
CAS 1219805-39-6
Molekylær formel C8 D3 H13 O2
Mængde 0.5 g
Synonym Hexyl alcohol acetate-d3, 1-Hexyl acetate-2,2,2-d3, Hexyl ethanoate-2,2,2-d3, n-Hexyl acetate-d3, Acetic acid-d3 hexyl ester
InChI formel InChI=1 S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3/i2D3
SMIL [2 H]C([2 H])([2 H])C(=O)OCCCCCC
IUPAC navn hexyl 2,2,2-trideuterioacetate
Molekylvægt (g/mol) 147.2299
Formel vægt 147.1339 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity