Få mere at vide
n-Hexyl-5,5,6,6,6-d5-triphenylphosphonium Bromide, CDN
Artikelnummer. 30312945
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312945 0.1 g
30314419 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312945 Leverandør CDN Leverandørnr. CDND78910.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexyltriphenylphosphonium-D5 (hexyl-5,5,6,6,6-D5) Bromide
CAS 1169764-53-7
Molekylær formel C24 D5 H23 P . Br
Mængde 0.1 g
Synonym Phosphonium, hexyl-5,5,6,6,6-d5-triphenyl-, bromide (1:1) (ACI), n-Hexyl-5,5,6,6,6-d5-triphenylphosphonium bromide, (Hexyl-5,5,6,6,6-d5)triphenylphosphonium bromide, Hexyltriphenylphosphonium bromide-d5, Hexyltriphenylphosphonium-D5 (hexyl-5,5,6,6,6-D5) bromide
InChI formel InChI=1 S/C24H28P.BrH/c1-2-3-4-14-21-25(22-15-8-5-9-16-22,23-17-10-6-11-18-23)24-19-12-7-13-20-24;/h5-13,15-20 H,2-4,14,21H2,1H3;1 H/q+1;/p-1/i1D3,2D2;
SMIL [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])CCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC navn 5,5,6,6,6-pentadeuteriohexyl(triphenyl)phosphanium;bromide
Molekylvægt (g/mol) 432.388
Formel vægt 431.143 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity