Få mere at vide
n-Hexyl-5,5,6,6,6-d5 Alcohol, TRC
Artikelnummer. 30179356
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179356 10 mg
30178933 1 mg
30179353 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179356 Leverandør Toronto Research Chemicals Leverandørnr. TRCH28122410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64118-18-9
Molekylær formel C6H9D5O
Molekylvægt (g/mol) 107.21
Synonym 1-Hexan-5,5,6,6,6-d5-ol (9CI), Hexyl-5,5,6,6,6-d5 alcohol, 1-Hexanol-d5, 1-Hexanol-5,5,6,6,6-D5, n-Hexyl-5,5,6,6,6-d5 alcohol, 1-Hexanol-5,5,6,6,6-d5
IUPAC navn hexan-5,5,6,6,6-d5-1-ol
SMIL [2H]C([2H])([2H])C([2H])([2H])CCCCO
Tekniske data
Kemisk navn eller materiale 1-Hexanol-5,5,6,6,6-D5
Mængde 10 mg
InChI formel InChI=1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2
Formel vægt 107.14