Få mere at vide
n-Hexyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Artikelnummer. 30324556
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324556 0.1 g 0.1g
30310766 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324556 Leverandør CDN Leverandørnr. CDND75860.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexyl 4-Hydroxybenzoate-2,3,5,6-D4
CAS 2241800-08-6
Molekylær formel C13 D4 H14 O3
Mængde 0.1 g
Synonym p-Hydroxybenzoic acid hexyl ester-d4, 4-Hydroxybenzoic acid-d4 hexyl ester, Hexyl 4-hydroxybenzoate-2,3,5,6-D4, Hexyl p-hydroxybenzoate-d4, Hexylparaben-d4, n-Hexyl 4-hydroxybenzoate-2,3,5,6-d4
InChI formel InChI=1 S/C13H18O3/c1-2-3-4-5-10-16-13(15)11-6-8-12(14)9-7-11/h6-9,14 H,2-5,10H2,1H3/i6D,7 D,8 D,9 D
SMIL [2 H]c1c([2 H])c(C(=O)OCCCCCC)c([2 H])c([2 H])c1O
IUPAC navn hexyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 226.305
Formel vægt 226.151 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity