Få mere at vide
n-Hexadecylpyridinium-d5 Bromide, TRC
Artikelnummer. 30470494
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470494 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470494 Leverandør Toronto Research Chemicals Leverandørnr. TRCN21125110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cetylpyridinium-D5 (pyridinium-D5) Bromide
CAS 143715-91-7
Molekylær formel C21H33D5N . Br
Mængde 10 mg
Synonym Pyridinium-2,3,4,5,6-d5, 1-hexadecyl-, bromide (9CI), Cetylpyridinium-D5 (pyridinium-D5) bromide, 1-Hexadecyl-2,3,4,5,6-pentadeuteriopyridinium bromide, 1-Cetylpyridinium-d5 bromide, Cetylpyridinium-d5 bromide, Hexadecylpyridinium-d5 bromide, n-Hexadecylpy
InChI formel InChI=1S/C21H38N.BrH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H/q+1;/p-1/i15D,17D,18D,20D,21D;
SMIL CCCCCCCCCCCCCCCC[N+](C([2H])=C1[2H])=C(C([2H])=C1[2H])[2H].[Br-]
IUPAC navn 1-hexadecylpyridin-1-ium-2,3,4,5,6-d5 bromide
Molekylvægt (g/mol) 389.48
Formel vægt 388.25