Få mere at vide
n-Hexadecyl-1,1,2,2-d4 Hexadecanoate-16,16,16-d3, CDN
Artikelnummer. 30326917
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326917 0.1 g
30315582 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326917 Leverandør CDN Leverandørnr. CDND73690.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexadecyl-1,1,2,2-D4 Hexadecanoate-16,16,16-D3
CAS 1398066-15-3
Molekylær formel C32 D7 H57 O2
Mængde 0.1 g
Synonym Cetyl-α,β-d4 palmitate-ω-d3, 1-Hexadecanol-1,1,2,2-d4 palmitate-16,16,16-d3, Palmitic acid cetyl ester-d7, Palmitic acid-d3 hexadecyl ester-d4, Hexadecyl-1,1,2,2-D4 hexadecanoate-16,16,16-D3, n-Hexadecyl-1,1,2,2-d4 hexadecanoate-16,16,16-d3
InChI formel InChI=1 S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3/i2D3,29D2,31D2
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCC(=O)OC([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCCCCCC
IUPAC navn 1,1,2,2-tetradeuteriohexadecyl 16,16,16-trideuteriohexadecanoate
Molekylvægt (g/mol) 487.892
Formel vægt 487.535 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity