Få mere at vide
n-Hexadecyl-1,1,2,2-d4 Alcohol, CDN
Artikelnummer. 30326911
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326911 0.5 g 0.5g
30323671 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326911 Leverandør CDN Leverandørnr. CDND73700.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Hexadecanol-1,1,2,2-D4
CAS 1398065-49-0
Molekylær formel C16 D4 H30 O
Mængde 0.5 g
Synonym 1-Hexadecan-1,1,2,2-d4-ol, [1,1,2,2-2H4]Hexadecanol, n-Hexadecanol-1,1,2,2-d4, n-Hexadecyl-1,1,2,2-d4 alcohol, Cetyl alcohol-α,β-d4, Cetyl-1,1,2,2-d4 alcohol, 1-Hexadecanol-1,1,2,2-D4
InChI formel InChI=1 S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3/i15D2,16D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])CCCCCCCCCCCCCC
IUPAC navn 1,1,2,2-tetradeuteriohexadecan-1-ol
Molekylvægt (g/mol) 246.465
Formel vægt 246.286 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity