Få mere at vide
N-Hexadecanoyl-2-amino-1,3-propane-d5-diol, CDN
Artikelnummer. 30328223
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328223 0.05 g -
30308885 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328223 Leverandør CDN Leverandørnr. CDND81070.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[2-Hydroxy-1-(hydroxymethyl)ethyl]hexadecanamide-D5 (2-aminopropane-1,3-diol-D5)
CAS 946524-35-2
Molekylær formel C19 D5 H34 N O3
Mængde 0.05 g
Synonym Hexadecanamide, N-[2-hydroxy-1-(hydroxymethyl-d2)ethyl-1,2,2-d3]- (ACI), N-[2-Hydroxy-1-(hydroxymethyl-d2)ethyl-1,2,2-d3]hexadecanamide (ACI), N-[2-Hydroxy-1-(hydroxymethyl)ethyl]hexadecanamide-D5 (2-aminopropane-1,3-diol-D5), Palmitoyl serinol-d5, N-Hexadecanoyl-2-amino-1,3-propane-d5-diol
InChI formel InChI=1 S/C19H39NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(23)20-18(16-21)17-22/h18,21-22 H,2-17H2,1H3,(H,20,23)/i16D2,17D2,18 D
SMIL [2 H]C([2 H])(O)C([2 H])(NC(=O)CCCCCCCCCCCCCCC)C([2 H])([2 H])O
IUPAC navn N-(1,1,2,3,3-pentadeuterio-1,3-dihydroxypropan-2-yl)hexadecanamide
Molekylvægt (g/mol) 334.549
Formel vægt 334.324 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity